Documents

BCR353

ZnAl4 (trace elements)

BCR®, certified reference material, disc

Synonym(s):
Aluminum-zinc alloy
CAS Number:
MDL number:

grade

certified reference material

Agency/Method

BCR®

form

disc

manufacturer/tradename

JRC

Featured Industry

Industrial Hygiene
Pharmaceutical (small molecule)

format

matrix material

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR353

Legal Information

BCR is a registered trademark of European Commission
Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.