CRM015

Supelco

Trace Metals - Fresh Water Sediment 2

pkg of 50 g

NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

grade

certified reference material

CofA

current certificate can be downloaded

packaging

pkg of 50 g

mfr. no.

RTC, CRM015-050

application(s)

AAS: suitable
ICP: suitable

Featured Industry

Environmental

format

matrix material

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

View example COA for CRM015

Note - The link provided is an example certificate of analysis (COA) only and may not be the lot that you receive. All analytes may not be present or certified for each lot, and analyte concentrations will vary by lot.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.
The following elements are not certified and are included for information only:
  • Sb, B, Cd, Mo, Se, Si, Ag, Sr, Tl

Analyte

Description

    Aluminum
    Antimony
    Arsenic
    Barium
    Beryllium
    Boron
    Cadmium
    Calcium
    Chromium
    Cobalt
    Copper
    Iron
    Lead
    Magnesium
    Manganese
    Mercury
    Molybdenum
    Nickel
    Potassium
    Selenium
    Silicon
    Sodium
    Strontium
    Thallium
    Vanadium
    Zinc
    See All (26)

Signal Word

Danger

Hazard Statements

RIDADR

UN 3077 9 / PGIII

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.