DEN15003

Density Standard (150 °C)

UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.8277 g/mL at 150 °C

NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

shelf life

limited shelf life, expiry date on the label

mfr. no.

Paragon Scientific Ltd

density

0.8277 g/mL at 150 °C

Featured Industry

Cleaning Products
Cosmetics
Flavors and Fragrances
Food and Beverages
Personal Care

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Density Standards can be found on the Paragon Scientific webpage

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.