Skip to Content
MilliporeSigma

LCL30

Lithium chloride solution

2 M in ethanol

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Linear Formula:
LiCl
CAS Number:
Molecular Weight:
42.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

concentration

2 M in ethanol

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

[Li+].[Cl-]

InChI

1S/ClH.Li/h1H;/q;+1/p-1

InChI key

KWGKDLIKAYFUFQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Lithium chloride may be used as a salt precursor for the preparation of stock solution of lithium for the determination of lithium ions in environmental water samples by ion chromatography equipped with a high-capacity cation-exchange column.[1] It may be used to generate lithium adducts of neutral carbohydrates present in food and beverage samples[2] and plants[3] so as to increase the sensitivity of detection for quadrupole mass spectrometry analysis (MS) with electrospray ionization (ESI) post their separation using anion exchange chromatography.

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

57.2 °F

flash_point_c

14 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of carbohydrates in plants by high-performance anion-exchange chromatography coupled with electrospray mass spectrometry
Guignard C, et al.
Journal of Chromatography A, 1085(1), 137-142 (2005)
Determination of inorganic cations and ammonium in environmental waters by ion chromatography with a high-capacity cation-exchange column
Thomas DH, et al.
Journal of Chromatography A, 956(1-2), 181-186 (2002)
Analysis of carbohydrates by anion exchange chromatography and mass spectrometry
Bruggink C, et al.
Journal of Chromatography A, 1085(1), 104-109 (2005)
Karla M Acevedo et al.
The Journal of biological chemistry, 289(16), 11007-11019 (2014-03-13)
Amyloid precursor protein (APP) undergoes post-translational modification, including O- and N-glycosylation, ubiquitination, and phosphorylation as it traffics through the secretory pathway. We have previously reported that copper promotes a change in the cellular localization of APP. We now report that
Ted M Hsu et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(2), 327-337 (2014-07-19)
Glucagon-like peptide-1 (GLP-1) is produced in the small intestines and in nucleus tractus solitarius (NTS) neurons. Activation of central GLP-1 receptors (GLP-1Rs) reduces feeding and body weight. The neural circuits mediating these effects are only partially understood. Here we investigate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service