LI0691BC

Millipore

TRANSIA PLATE Listeria spp.

BioControl, ELISA test for detection of Listeria in food and environmental samples

Agency/Method

AFNOR

shelf life

3‑24 mo.

packaging

pkg of 96 tests

manufacturer/tradename

BioControl Systems LI0691
TRANSIA

application(s)

ELISA: suitable
pathogen testing: suitable

input

food(s)

Featured Industry

Environmental
Food and Beverages

storage temp.

2-8°C

suitability

Listeria spp.

General description

TRANSIA PLATE Listeria is a kit for detection of the Listeria genus(L. monocytogenes, L. ivanovii, L. innocua, L. seeligeri, L. welshimeri, L. grayi )in dairy products, meat products, seafood products, vegetables, and environmental samples.

Application

AFNOR Certification TRA 02/06 – 11/95

Features and Benefits

  • Detects all 6 species of Listeria providing a true indication of product and surface quality.
  • Positive and Negative controls supplied with the kit.

Components

  • 1 microplate with dividable microwell strips with 96 wells (12 microwell strips of 8 microwells).
  • Negative control: (bacterial antigens not belonging to the Listeria genus), ready-to-use, 10 mL.
  • Positive control: Listeria monocytogenes antigens, ready-to-use, 8 mL.
  • Conjugate (contains peroxidase conjugated anti-Listeria antibodies), ready-to-use, 16 mL.
  • Wash buffer, 20 X concentration, 60 mL.
  • Substrate (urea H2O2), ready-to-use, 57 mL.
  • Chromogen (TMB), ready-to-use, 57 mL.
  • Stop solution (H2SO4), ready-to-use, 57 mL.
  • 1 microplate cover.

Principle

TRANSIA PLATE Listeria is an ELISA test (Enzyme Linked Immuno Sorbent Assay) based on a two step sandwich-type reaction. The solid support of the reaction is a microtitration plate with dividable microwellstrips. Specific antibodies of flagellar antigens are grafted on the microtitration plate wells.

Testing Method

ELISA

Legal Information

TRANSIA is a trademark of BioControl Systems, Inc.

Kit Components Also Available Separately

Product No.
Description
SDS

  • Wash Buffer, 20X, 60 mL, 96 test

  • Substrate, 57 mL

  • Chromogen, 57 mL, 960 test

  • Stop Solution, 57 mL, 960 test

  • Coated Plates, 96 wells

  • Conjugate, 16 mL, 96 test

  • Positive Control, 8 mL Fill

  • Negative Control, 10 mL Fill

See All (8)

Signal Word

Danger

hazcat

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 1

storage_class_code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Related Content
TRANSIA AG (Enzyme ImmunoAssay) and TRANSIA® PLATE incorporate proven antibody-antigen "sandwich" assay technology for the detection of food borne pathogens in both product and environmental samples
Read More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service