M100

Supelco

(−)-N-Methylphysostigmine

analytical standard, for drug analysis

Empirical Formula (Hill Notation):
C16H23N3O2
CAS Number:
Molecular Weight:
289.37
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard, for drug analysis

form

solid

optical activity

[α]25/D −67.6°, c = 1 in chloroform(lit.)

application(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

solubility

H2O: 1.9 mg/mL
DMSO: 13 mg/mL
ethanol: soluble
methanol: soluble

Featured Industry

Forensics and Toxicology
Veterinary

format

neat

storage temp.

2-8°C

SMILES string

CN(C)C(=O)Oc1ccc2N(C)[C@H]3N(C)CC[C@@]3(C)c2c1

InChI

1S/C16H23N3O2/c1-16-8-9-18(4)14(16)19(5)13-7-6-11(10-12(13)16)21-15(20)17(2)3/h6-7,10,14H,8-9H2,1-5H3/t14-,16+/m1/s1

InChI key

IUHMWLMDASQDJQ-ZBFHGGJFSA-N

Gene Information

human ... ACHE(43), BCHE(590)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Hygroscopic

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 1544PSN1 6.1 / PGII

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Gary H Chang et al.
Scientific reports, 9(1), 6839-6839 (2019-05-03)
The hallmark of drug-coated balloon (DCB) therapy for the treatment of peripheral vascular disease is that it allows for reopening of the narrowed lumen and local drug delivery without the need for a permanent indwelling metal implant such as a...
Claudio Ferrante et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 125, 452-461 (2019-02-04)
One of the most promising economic perspectives of hemp production chain is female inflorescence valorization, despite there being actually no chemical composition or biological data from water fraction. In this context, the focus of this study is the evaluation of...
Maura Gasparetto et al.
Leukemia research, 80, 1-10 (2019-03-11)
Iron metabolism is altered in a variety of cancers; however, little is known about the role of iron metabolism in the biology and response to therapy of acute myeloid leukemia (AML). Here we show that SLC40A1, the gene encoding the...
Petya Dimitrova et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 55, 105-118 (2019-01-23)
Neutrophils have a short live in circulation and accelerate greatly local immune responses via increased granulopoiesis and migration at high numbers to infected or inflamed tissue. Since neutrophils produce a variety of factors with destructive and pro-inflammatory potential the regulation...
Hira Butt et al.
European journal of pharmacology, 872, 172958-172958 (2020-02-01)
The deteriorating effects of heat stress abrogate the therapeutic implications of human Wharton's jelly derived mesenchymal stem cells (hWJMSCs) transplanted in burn wounds. Topically applied green tea extract comprising epigallocatechin-3-gallate (EGCG) is known to repair burn wounds. Here, we investigated...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.