NIST2296

Reformulated gasoline (nominal mass fraction 13 % ETBE)

NIST® SRM® 2296

NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

grade

certified reference material

packaging

pkg of 2 x 20 mL

mfr. no.

NIST

Featured Industry

Pharmaceutical (small molecule)

format

matrix material

General description

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Signal Word

Danger

Target Organs

Central nervous system

RIDADR

UN1203 - class 3 - PG 2 - Gasoline

WGK Germany

WGK 3

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.