Skip to Content
MilliporeSigma

P7971

Potassium alum dodecahydrate

meets USP testing specifications

Synonym(s):

Aluminum potassium sulfate dodecahydrate, Alum, Potassium alum, Potassium aluminum sulfate dodecahydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
AlK(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
474.39
UNSPSC Code:
12352200
PubChem Substance ID:
EC Number:
233-141-3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

meets USP testing specifications

mp

92 °C (lit.)

density

1.757 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Al.K.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;;2*(H2,1,2,3,4);12*1H2/q+3;+1;;;;;;;;;;;;;;/p-4

InChI key

GNHOJBNSNUXZQA-UHFFFAOYSA-J



PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library