MilliporeSigma
All Photos(1)

PFS49

Supelco

SpheriCal Mix DHB

Protein Medium

form

powder

shelf life

expiry date on the label

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

SpheriCal is a dendrimer-based MALDI calibration kit providing 4 monodisperse mass signals per kit for efficient, highly accurate calibration. By combining excellent long-term stability with evenly-spaced calibration points, SpheriCal helps users increase the frequency and quality of their MALDI-MS calibrations. With a shelf life of years and each kit offering at least 100 calibration spots, SpheriCal offers a reliable and accurate calibration across a large mass range, without the shelf-life and storage issues associated with protein or peptide-based systems.

Mass ranges available:

i.SpheriCal Mix DHB Peptide Low(m/z 500 - 1,600)
ii.SpheriCal Mix DHB Peptide Medium(m/z 1,600 - 3,500)
iii.SpheriCal Mix DHB Peptide High(m/z 3,500 - 7,500)
iv.SpheriCal Mix DHB Protein Low (m/z 7,500 - 15,000)
v.SpheriCal DHB Protein Medium (m/z 15,000 - 30,000)

SpheriCal Mix: quick and easy to use - just add solvent

As calibration standard, matrix and counter ion are pre-mixed in optimized ratios, SpheriCal Mix dramatically expedites sample preparation for calibration in minutes. SpheriCal Mix DHB is pre-formulated with dihydroxybenzoic acid, a commonly used matrix in the analysis of peptides, proteins, oligosaccharides, nucleotides and synthetic polymers.

Find more information here SpheriCal

Legal Information

SpheriCal is a trademark of Polymer Factory Sweden AB

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service