Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day through March 27.Save Now

PHR3375

Triclocarban Related Compound A

certified reference material, pharmaceutical secondary standard

Synonym(s):

1,3-Bis(4-chlorophenyl)urea, 4,4′-Dichlorocarbanilide

Sign In to View Organizational & Contract Pricing.

Select a Size

30 MG

$492.00

$492.00


Check Cart for Availability

Request a Bulk Order

About This Item

Empirical Formula (Hill Notation):
C13H10Cl2N2O
CAS Number:
Molecular Weight:
281.14
UNSPSC Code:
41116107
NACRES:
NA.24

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

grade

certified reference material, pharmaceutical secondary standard

agency

traceable to USP 1681815

packaging

pkg of 30 mg

application(s)

pharmaceutical

storage temp.

2-30°C

SMILES string

Clc1ccc(cc1)NC(=O)Nc2ccc(cc2)Cl

InChI

1S/C13H10Cl2N2O/c14-9-1-5-11(6-2-9)16-13(18)17-12-7-3-10(15)4-8-12/h1-8H,(H2,16,17,18)

InChI key

ZNQCSLYENQIUMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PHR3149PHR3297PHR2915
grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

pharmaceutical

agency

traceable to USP 1681815

agency

traceable to USP 1681845

agency

traceable to USP 1150513

agency

traceable to USP 1119309

packaging

pkg of 30 mg

packaging

pkg of 40 mg

packaging

pkg of 50 mg

packaging

pkg of 50 mg

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

storage temp.

2-8°C

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP and EP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

comparable product

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
PHR3375-30MG04065267400716

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service