Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR8886

Supelco

Triacetylclindamycin

pharmaceutical secondary standard, certified reference material

Synonym(s):

[(2R,3S,4S,5R,6R)=3,5-diacetyloxy-2-[(1S,2S)-2-chloro-1-[[(2S,4R)-1-methyl-4-propylpyrrolidine-2-carbonyl]amino]propyl]-6-methylsulfanyloxan-4-yl] acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H39ClN2O8S
Molecular Weight:
551.09
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

CofA

current certificate can be downloaded

packaging

pkg of 100 mg

application(s)

pharmaceutical

storage temp.

-10 to -25°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PHR3709PHR2059PHR3720
application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

pharmaceutical

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

packaging

pkg of 100 mg

packaging

pkg of 100 mg

packaging

pkg of 25 mg

packaging

pkg of 50 mg

storage temp.

-10 to -25°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Lact. - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service