Skip to Content
MilliporeSigma

Skip To

PMC1102

Acetonitrile

≥99.75%, ISOM8™, anhydrous

Synonym(s):

ACN, Acetonitrile, MeCN, Cyanomethane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL

Available to ship TODAY

$61.80
2 L

Available to ship TODAY

$185.00
4 x 2 L

Available to ship TODAY

$495.00
12 x 100 mL

Available to ship TODAY

$618.00

About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
UNSPSC Code:
12352148
Beilstein/REAXYS Number:
741857
MDL number:
Assay:
≥99.75%
Grade:
anhydrous
Bp:
81-82 °C (lit.), 82 °C ((lit))
Vapor pressure:
44 mmHg ( 20 °C), 72.8 mmHg ( 20 °C)

$61.80


Available to ship TODAYDetails


​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

anhydrous

vapor density

1.41 (vs air), 1.59 (vs air)

vapor pressure

44 mmHg ( 20 °C), 72.8 mmHg ( 20 °C)

description

Various Returnable Container Options

product line

ISOM8™

assay

≥99.75%

form

liquid

autoignition temp.

523 °C (975°F), 973 °F

shelf life

36 mo.

expl. lim.

16 %

packaging

Sure/Seal™ of 100 mL, Sure/Seal™ of 12 × 100 mL, Sure/Seal™ of 2 L, Sure/Seal™ of 4 × 2 L

impurities

<0.001% water (<0.005% water for individual packaging amounts less than 500 mL)

evapn. residue

<0.0005%

refractive index

n/D 1.3440 (lit.), n20/D 1.344 (lit.)

pH

(10 ppmacidity (format:neat))

bp

81-82 °C (lit.), 82 °C ((lit))

mp

-45 °C, −45 °C (lit.)

transition temp

flash point 6 °C (42°F)

solubility

miscible

density

0.786 g/mL at 25 °C

cation traces

(See Dossier)

application(s)

diagnostic assay manufacturing
life science and biopharma

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

General description

Welcome to our ISOM8™ line of high-quality products available and able to scale for manufacturing. Within our ISOM8 team, we have expertise and documentation to support ISO 9001 industrial and non-GMP pharma production needs, as defined by our M-Clarity™ Program.

Our portfolio of ISOM8 products and chemistries is ever evolving to support our valued Sigma-Aldrich® customers′ requirements. Please navigate to our documents section to look at dossiers and other quality documentation for this product, provided with support of our Elevated Assurance Program.

Our ISOM8™ Acetonitrile is available within the USA with our ReCycler® Solvent System for returnable, reusable, and readily available bulk solvent delivery, to reduce packaging waste and help you achieve your sustainablility goals. To request ISOM8 and other product lines in returnable containers Click Here.

Unlock your manufacturing potential with our comprehensive program. By choosing ISOM8™ chemicals, ensure compliance while fostering innovation and excellence. Connect with us to learn more about how we can support your journey toward manufacturing success.

Application

Organic Synthesis
Acetonitrile plays a crucial role in various organic reactions, including nucleophilic substitutions and coupling reactions, facilitating the formation of complex molecules.
Extraction Processes
This solvent is effective for extracting active ingredients and impurities from raw materials, enhancing the efficiency of purification processes in both industrial and non-GMP pharmaceutical settings.

Features and Benefits

FEATURES


  • Anhydrous Solvent: The absence of water ensures that it is ideal for reactions and processes that require moisture-free conditions.
  • High Purity: We ensure that our anhydrous acetonitrile meets stringent quality standards, making it suitable for a wide range of demanding applications.
  • Versatile Applications: Acetonitrile is a polar aprotic solvent, which makes it effective for a wide range of chemical reactions, including nucleophilic substitutions and extractions.
  • Returnable Containers: Our anhydrous acetonitrile is offered in returnable containers, designed for industry to minimize waste, and support sustainable practices.
BENEFITS


  • Improved Reaction Efficiency: The anhydrous nature reduces side reactions that can occur in the presence of water, possibly improving yields in select chemical syntheses.
  • Cost Efficiency: Comparably lower-cost, non-GMP product production combined with returnable containers can lead to significant cost savings over time.
  • Convenience: The returnable container system simplifies logistics to manage inventory and replenishment without the need for additional waste disposal.

Packaging

100mL to 2L bottles are packaged in Sure/Seal™containers.

For bulk amounts in US, we offer returnables through ReCycler® Program.

Legal Information

ISOM8 is a trademark of Sigma-Aldrich Co. LLC
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC
M-CLARITY is a trademark of Merck KGaA, Darmstadt, Germany
RECYCLER is a registered trademark of Merck KGaA, Darmstadt, Germany
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AX015234442L010002
description

≥99.75%, ISOM8™, anhydrous

description

anhydrous, Biotechnology, ≥99.9% (GC)

description

suitable for DNA synthesis, ≥99.9% (GC)

description

-

grade

anhydrous

grade

biotech. grade, anhydrous

grade

-

grade

-

assay

≥99.75%

assay

≥99.9% (GC)

assay

≥99.9% (GC)

assay

-

Quality Level

300

Quality Level

300, 200

Quality Level

100

Quality Level

200

application(s)

diagnostic assay manufacturing
life science and biopharma

application(s)

pharmaceutical

application(s)

-

application(s)

-

solubility

miscible

solubility

-

solubility

-

solubility

water: soluble (completely)

form

liquid

form

liquid

form

liquid

form

liquid


Still not finding the right product?

Explore all of our products under Acetonitrile


Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

Flash Point (°F)

35.6 °F - closed cup

Flash Point (°C)

2.0 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library