SA0180BC

Millipore

TRANSIA PLATE Salmonella Gold

BioControl, ELISA test for detection of Salmonella in food and environmental samples

Synonym(s):
TPSG

Agency/Method

AOAC® (PTM)

shelf life

3‑9 mo.

packaging

pkg of 96 tests

manufacturer/tradename

BioControl Systems SA0180
TRANSIA

application(s)

ELISA: suitable
pathogen testing: suitable

input

food(s)

Featured Industry

Environmental
Food and Beverages

storage temp.

2-8°C

suitability

Salmonella spp.

General description

TRANSIA PLATE Salmonella Gold is an ELISA kit intended to be used by food microbiologists for the detection of Salmonella genus. The assay reliably recovers and detects low levels of Salmonella in a variety of foods and feeds.

Application

AFNOR Certification TRA 02/08 – 03/01, AOAC Performance Tested Method 010602

Features and Benefits

  • Accurate Method for Salmonella Detection:

TRANSIA PLATE Salmonella Gold is a sandwich enzyme immunoassay for the detection of Salmonella in food, feed and environmental samples. The test utilizes highly specific proprietary antibodies to create an antigen antibody complex which, if present, creates a color change reaction upon addition of the substrate which is read with a microplate reader.

  • Standardized Protocol – Results in 24 hours:

The same enrichment media and incubation time for all samples reduces media preparation time and increases lab efficiency. Enrichment steps are harmonized with ISO reference method and are common for all matrices.

  • Automation Ready:

TRANSIA Plate Salmonella Gold can be fully automated with the Gemini® microplate processor.

  • Extensively validated


TRANSIA Plate Salmonella Gold has numerous validations covering all foods, feed and environmental samples from organizations including AFNOR and AOAC Research Institute.

Components

  • Microtiter plate with divisible strips, 96 wells (8 wells x 12 strips) – 1 pc
  • Lid for the microtiter plate – 1 pc
  • Negative control – ready to use – 1 x 10 mL
  • Positive control – ready to use – 1 x 8 mL
  • Washing buffer – concentrated 20X – 1 x 60 mL
  • Conjugate: anti-Salmonella antibodies conjugated to peroxydase – ready to use – 1 x 16 mL
  • Substrate: TMB – ready to use – 1 x 57 mL
  • Stop solution: H2SO4 – ready to use – 1 x 57 mL
  • Extraction reagent – ready to use – 1 x 12.5 mL

Testing Method

ELISA

Legal Information

AOAC is a registered trademark of AOAC International
Gemini is a registered trademark of Stratec
TRANSIA is a trademark of BioControl Systems, Inc.

Kit Components Also Available Separately

Product No.
Description
SDS

  • Transia Plate Salmonella Gold Substrate, Filled 57 mL

  • Transia Plate Extraction Reagent, Filled 12.5 mL, 100 test

  • Wash Buffer, 20X, 60 mL, 96 test

  • Stop Solution, 57 mL, 960 test

  • Transia Plate Salmonella Gold Coated Plate

  • Transia Plate Salmonella Gold Conjugate, Filled 16 mL

  • Transia Plate Salmonella Gold Positive Control, Filled 8 mL

  • Transia Plate Salmonella Gold Negative Control, Filled 10 mL

See All (8)

Signal Word

Danger

hazcat

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

storage_class_code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Related Content
TRANSIA AG (Enzyme ImmunoAssay) and TRANSIA® PLATE incorporate proven antibody-antigen "sandwich" assay technology for the detection of food borne pathogens in both product and environmental samples
Read More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service