SPE018

Supelco

Phenols - PT

Proficiency Testing Material

EC Number:
NACRES:
NA.24

Quality Level

packaging

pkg of 1 × 40 g

matrix

soil

Featured Industry

Environmental

format

matrix material

General description

Phenols - PT is produced following the ISO/IEC 17043:2010. The PT portfolio allows our test components to offer significant performance data to monitor and prove competency to customers, accreditation bodies or both.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. We are the world leader in the manufacturing of real-world soil and sediment PT products.

Type of PT Scheme:
Solids/Soils/Hazardous Waste

Matrix: Soil

Analyte/s:
Concentrations may vary and not all analytes may be present

All information regarding the use of Phenols Proficiency Testing standard can be found in the reporting packet which will be available to download at PT Portal.

Application

Phenols proficiency testing material is widely used for environmental testing in Solids/Soils/Hazardous Waste.

Have any questions? Explore our FAQs.
For more information on this product, PT Service Team..

Preparation Note

Description

Randomly contains a minimum 80% of the analytes listed.

  • This PT sample is a soil containing extractable phenols for analysis by 8041 and/or 8270.
  • This sample consists of 10 g of soil.
  • Four samples have been provided for your convenience (multiple methods, multiple analysts, etc.)
  • The soil has been chemically stabilized to minimize degradation of the sample.
  • The PT sample has been sized to 100 mesh.
  • The evaluation of results will be based on study statistics as required by TNI.
  • The sample has been intentionally prepared with an apparent headspace.

Sample Preparation


  • Extract the complete contents of a single vial.
  • Transfer the entire contents of one vial to an extraction vessel.
  • Rinse vial and cap with extraction solvent.
  • Note: Sample extracts and calibration solutions should be in the same solvent.
  • Analyze the extract using the appropriate Semi-Volatile Organics method for which you are seeking accreditation.
  • Determine the concentration of the analytes listed and report your results on the organic analysis data sheet provided. The method used must be reported. If multiple methods are used for preparation and/or analysis, a separate report form, must be used for each preparation and analysis method.
  • Report all results on a wet weight basis.
  • Do not correct for moisture.
  • Assume a 10 g sample size for all calculations.

Storage and Stability

Please refer to sample label for expiration date, specific storage temperature, and specific storage conditions.

Other Notes

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ¢your reference¢ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Concentrations may vary and not all analytes may be present.

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes.

Proficiency Testing Schedule (2021-2022)

Soils
LPTP21-S2April 7 - May 21, 2021

LPTP22-S2April 6 - May 20, 2022

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    4-Chloro-3-methylphenol 500-15000 μg/kg
    2-Chlorophenol 500-15000 μg/kg
    2,4-Dichlorophenol 500-15000 μg/kg
    2,6-Dichlorophenol 500-15000 μg/kg
    2,4-Dimethylphenol 500-15000 μg/kg
    2,4-Dinitrophenol 500-15000 μg/kg
    2-Methyl-4,6-dinitrophenol 500-15000 μg/kg
    2-Methylphenol (o-Cresol) 500-15000 μg/kg
    3-Methylphenol (m-Cresol) 500-15000 μg/kg
    4-Methylphenol (p-Cresol) 500-15000 μg/kg
    3+4-Methylphenol (m+p-Cresol) 500-15000 μg/kg
    2-Nitrophenol 500-15000 μg/kg
    4-Nitrophenol 500-15000 μg/kg
    Pentachlorophenol 500-15000 μg/kg
    Phenol 500-15000 μg/kg
    2-Sec-butyl-4,6-dinitrophenol (DNBP, Dinoseb) 500-15000 μg/kg
    2,3,4,5-Tetrachlorophenol 500-15000 μg/kg
    2,3,4,6-Tetrachlorophenol 500-15000 μg/kg
    2,3,5,6-Tetrachlorophenol 500-15000 μg/kg
    2,4,5-Trichlorophenol 500-15000 μg/kg
    2,4,6-Trichlorophenol 500-15000 μg/kg
    See All (21)

RIDADR

NONH for all modes of transport

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.