SPE022

Supelco

Nitrosamines/Nitroaromatics - PT

Proficiency Testing Material

EC Number:
NACRES:
NA.24

Quality Level

packaging

pkg of 1 × 30 g

matrix

soil

Featured Industry

Environmental

format

matrix material

storage temp.

2-8°C

General description

Nitrosamines/Nitroaromatics - PT proficiency testing material is produced in accordance with ISO/IEC 17043:2010.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. We are the world leader in the manufacturing of real-world soil and sediment PT products. The PT portfolio allows our test components to offer significant performance data to monitor and prove competency to customers, accreditation bodies or both.

Type of PT Scheme:
Solids/Soils Hazardous Waste

Matrix:
Soil

Analyte/s: Values of analytes vary lot to lot.

All information regarding the use of Nitrosamines/Nitroaromatics - PT can be found in the reporting packet which will be available to download at PT Portal.

Application

Nitrosamines/Nitroaromatics - PT is a soils/hazardous waste proficiency testing material, suitable for testing explosives and explosive residue analytes in soil samples.

Our soil PT samples behave as real samples; and, unless there is no alternative, are not simply spikes added to a blank soil. They are designed to meet the requirements of national, regional and state-specific accreditation bodies (TNI, USEPA, UKAS) for quality assurance and continuous improvement.

Have any questions? Explore our FAQs.
For more information on this product, please contact our PT Service Team.

Features and Benefits

We have a vast portfolio of environmental PT schemes with "fit-for-purpose" samples to help you meet the needs of the laboratory.

Our proficiency testing (PT) program is a benchmark to:

  • provide staff with an insight into their laboratory′s performance, to verify the accuracy and reliability of their testing process
  • provide a comparison of performance with that of other laboratories, nationally and internationally
  • test and educate staff, provide a better understanding of the impact of incorrect results and identify areas where there may be problems
  • provide an independent evaluation of laboratory performance
  • demonstrate to clients, colleagues and accreditation bodies that there is a commitment to quality

Preparation Note

Description

  • The PT sample is a soil containing trace levels of explosives and explosive residues.
  • The PT sample size provided is 30 g of soil.
  • The soil has been chemically stabilized to minimize degradation of the sample.
  • The PT sample has been sized to 100 mesh.
  • Required storage condition is 4º C.
  • The sample has been intentionally prepared with an apparent headspace.

Sample Preparation
  • Analyze the sample using the preparatory and determinative method(s) for which you are seeking accreditation. The sample was specifically designed for SW846 8330, but is suitable for equivalent methods. Nitroglycerin must be analyzed using method 8332.
  • If you are accrediting for more than one method make additional copies of the report form and clearly indicate the method used for the data being reported.
  • Determine the concentration and report your results and method on the analysis data sheet provided.
  • Results are to be reported on a wet weight basis.

Storage and Stability

Please refer to sample label for expiration date, specific storage temperature, and specific storage conditions.

Other Notes

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ′your reference′ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes.

Proficiency Testing Schedule (2021-2022)

Soils
LPTP21-S2April 7 - May 21, 2021

LPTP22-S2April 6 - May 20, 2022

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    4-Amino-2,6-dinitrotoluene (4-am-dnt) 1500-15000 μg/kg
    2-Amino-4,6-dinitrotoluene (2-am-dnt) 1500-15000 μg/kg
    1,3-Dinitrobenzene (1,3-DNB) 1500-15000 μg/kg
    2,4-Dinitrotoluene (2,4-DNT) 1500-15000 μg/kg
    2,6-Dinitrotoluene (2,6-DNT) 1500-15000 μg/kg
    HMX (Octahydro-1,3,5,7-tetranitro-1,3,5,7-tetrazocine) 1500-15000 μg/kg
    Nitrobenzene 1500-15000 μg/kg
    Nitroglycerin 1500-15000 μg/kg
    2-Nitrotoluene 1500-15000 μg/kg
    3-Nitrotoluene 1500-15000 μg/kg
    4-Nitrotoluene 1500-15000 μg/kg
    Pentaerythritoltetranitrate (PETN) 1500-15000 μg/kg
    RDX (hexahydro-1,3,5-trinitro-1,3,5-triazine) 1500-15000 μg/kg
    Tetryl (Methyl-2,4,6-trinitrophenylnitramine) 1500-15000 μg/kg
    1,3,5-Trinitrobenzene (1,3,5-TNB) 1500-15000 μg/kg
    2,4,6-Trinitrotoluene (2,4,6-TNT) 1500-15000 μg/kg
    See All (16)

RIDADR

NONH for all modes of transport

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.