All Photos(1)

04270

Sigma-Aldrich

Ethylmalonic acid

purum, ≥97.0% (T)

Linear Formula:
C2H5CH(COOH)2
CAS Number:
Molecular Weight:
132.11
Beilstein:
774334
EC Number:
MDL number:
PubChem Substance ID:

grade

purum

assay

≥97.0% (T)

mp

110-114 °C
112-114 °C (lit.)

SMILES string

CCC(C(O)=O)C(O)=O

InChI

1S/C5H8O4/c1-2-3(4(6)7)5(8)9/h3H,2H2,1H3,(H,6,7)(H,8,9)

InChI key

UKFXDFUAPNAMPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Mainak Das et al.
Biomaterials, 30(29), 5392-5402 (2009-07-25)
The use of defined in vitro systems to study the developmental and physiological characteristics of a variety of cell types is increasing, due in large part to their ease of integration with tissue engineering, regenerative medicine, and high-throughput screening applications.
Carlo Viscomi et al.
Nature medicine, 16(8), 869-871 (2010-07-27)
Ethylmalonic encephalopathy is caused by mutations in ETHE1, a mitochondrial matrix sulfur dioxygenase, leading to failure to detoxify sulfide, a product of intestinal anaerobes and, in trace amounts, tissues. Metronidazole, a bactericide, or N-acetylcysteine, a precursor of sulfide-buffering glutathione, substantially
Patrícia Fernanda Schuck et al.
Neurochemical research, 35(2), 298-305 (2009-09-17)
High concentrations of ethylmalonic acid are found in tissues and biological fluids of patients affected by ethylmalonic encephalopathy, deficiency of short-chain acyl-CoA dehydrogenase activity and other illnesses characterized by developmental delay and neuromuscular symptoms. The pathophysiological mechanisms responsible for the
D I Zafeiriou et al.
Neuropediatrics, 38(2), 78-82 (2007-08-23)
Ethylmalonic encephalopathy (EE) is a rare, recently defined inborn error of metabolism which affects the brain, gastrointestinal system and peripheral blood vessels and is characterized by a unique constellation of clinical and biochemical features. A 7-month-old male, who presented with
M-H Read et al.
Journal of inherited metabolic disease, 28(6), 1149-1150 (2006-01-26)
We report the fourth case of combined D-and L-2-hydroxyglutaric aciduria presenting with neonatal encephalopathy and subependymal cysts.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service