MilliporeSigma
All Photos(1)

121S

Sigma-Aldrich

CD21 Positive Control Slides

suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections)

NACRES:
NA.24

packaging

pkg of 5 slides

Quality Level

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

shipped in

ambient

storage temp.

no temp limit

Compare Similar Items

View Full Comparison

1 of 4

This Item
115S171S144S
packaging

pkg of 5 slides

packaging

pkg of 5 slides

packaging

pkg of 5 slides

packaging

pkg of 5 slides

manufacturer/tradename

Cell Marque

manufacturer/tradename

Cell Marque

manufacturer/tradename

Cell Marque

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

no temp limit

storage temp.

no temp limit

storage temp.

no temp limit

storage temp.

no temp limit

Quality Level

100

Quality Level

100, 500

Quality Level

100, 500

Quality Level

100, 500

General description

CD21 positive control slides are intended for use as positive controls for immunohistochemical (IHC) staining using an anti-CD21 antibody.

Physical form

Paraffin embedded tissue section mounted on microscope slide

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service