19096

Sigma-Aldrich

Atto Rho11 Biotin

NACRES:
NA.32

Quality Level

fluorescence

λex 571 nm; λem 595 nm

storage temp.

−20°C

Application

Atto Rho11 Biotin is a new rhodamine dye for coupling to bio-molecules such as DNA, RNA, proteins etc. The label shows very high fluorescence efficiency and exceptionally high photostability. The dye is moderately hydrophilic. ATTO Rho11 is a cationic dye. After coupling to a substrate the label carries a net electrical charge of +1. Biotin conjugates can be used in applications like ELISA or immunohistochemistry, in situ hybridization, flow cytometry and others, to identify streptavidin, avidin, or extravidin-conjugates.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Rhodamine-inspired far-red to near-infrared dyes and their application as fluorescence probes.
Sun YQ, Liu J, Lv X, Liu Y, Zhao Y, Guo W.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 51, 7634-7636 (2012)
Jan Klohs et al.
Basic research in cardiology, 103(2), 144-151 (2008-03-08)
Light in the near-infrared (NIR) region between 700-900 nm can penetrate deep into living tissue, thereby offering a unique opportunity to use near-infrared fluorescence (NIRF) imaging techniques to detect and visualize fluorescent probes in-vivo. In the past few years, many...
Chunbai He et al.
Biomaterials, 33(33), 8569-8578 (2012-08-22)
Polymeric nanoparticles have been widely applied to oral delivery of protein drugs, however, few studies focused on the systematical elucidation of the size-dependent oral absorption mechanism with well-defined polymeric nanoparticles. Rhodamine B labeled carboxylated chitosan grafted nanoparticles (RhB-CCNP) with different...
Jianghong Rao et al.
Current opinion in biotechnology, 18(1), 17-25 (2007-01-20)
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals. To overcome the photon attenuation in living tissue, fluorophores with long emission at the near-infrared (NIR) region are generally preferred, including...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.