200S

Sigma-Aldrich

Androgen Receptor Positive Control Slides

suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections)

NACRES:
NA.24

Quality Level

packaging

pkg of 5 slides

manufacturer/tradename

Cell Marque

application(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

shipped in

ambient

storage temp.

2-8°C

General description

Androgen Receptor positive control slides are intended for use as positive controls for immunohistochemical (IHC) staining using an anti-Androgen Receptor antibody.

Physical form

Paraffin embedded tissue section mounted on microscope slide

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Sigma-Aldrich Co. LLC

storage_class_code

13 - Non Combustible Solids

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Yusuke Fugane et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 36(11), 1415-1421 (2020-07-28)
This paper describes uncertainty ranges of the analytical values by laser-induced breakdown spectrometry (LIBS), in order to realize an element-based sorting of commercial Al alloys whose chemical compositions are varied depending on the kind. For this purpose, calibration factors and...
Mariane Wolf et al.
International journal of biological macromolecules, 115, 157-164 (2018-04-15)
β-d-galactosidase was immobilized in a modified Arabic gum-based hydrogel for the hydrolysis of standard lactose and lactose contained in UHT milk with the aim of producing lactose-free/low-lactose milk. The hydrogel was synthesized by cross-linking the modified Arabic gum with acrylamide...
Denis Chateau et al.
ACS applied materials & interfaces, 11(42), 39068-39076 (2019-10-01)
Gold nanoparticles offer unique optoelectronic properties relevant for a wide range of processes and products, in biology and medicine (therapeutic agents, diagnostic, drug delivery), as well as in electronics, photovoltaics, and catalysis. So far, various synthesis methods proposed have led...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service