Skip to Content
MilliporeSigma

Skip To

50247

Humulone, mixture of homologues

≥90.0% (HPLC)

Synonym(s):

α-Bitter acid, α-Lupulic acid

Sign In to View Organizational & Contract Pricing.

About This Item

CAS Number:
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:


assay

≥90.0% (HPLC)

storage temp.

−20°C

SMILES string

CC(C)CC(=O)C1=C(O)[C@](O)(C\C=C(\C)C)C(=O)C(C\C=C(\C)C)=C1O

Analysis Note

predominantly humulone and cohumulone


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library