73948

Sigma-Aldrich

Atto 655 iodoacetamide

suitable for fluorescence

NACRES:
NA.32

fluorescence

λex 663 nm; λem 684 nm in 0.1 M phosphate pH 7.0

Quality Level

suitability

corresponds for coupling to thiols
suitable for fluorescence

storage temp.

−20°C

General description

Atto 655 belongs to a new generation of fluorescent labels. The dye is designed for application in the area of life science, e.g. labeling of DNA, RNA or proteins. Characteristic features of the label are strong absorption, good fluorescence quantum yield, excellent thermal and photo-stability, outstanding ozone resistance, very good water solubility, and very little triplet formation. Atto 655 is a zwitterionic dye with a net electrical charge of zero. The fluorescence is efficiently quenched by electron donors like guanine, tryptophan, etc.
The iodoacetamide derivative reacts, like the maleimide, with a sulfhydryl group forming a thioether bond. It is predominantly used for tagging cystein residues of proteins.

find more information
here

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Bengang Xing et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(50), 14170-14177 (2011-11-16)
The molecular interactions of the glycopeptide antibiotic vancomycin (Van) with bacterial cell wall analogues N,N'-diacetyl-L-Lys-D-Ala-D-Ala (Ac(2) KdAdA) and N,N'-diacetyl-L-Lys-D-Ala-D-Lac (Ac(2) KdAdL) were investigated in neat water, phosphate buffer and HEPES buffer by using fluorescence correlation spectroscopy (FCS) and molecular dynamics...
Fabian Heinemann et al.
Langmuir : the ACS journal of surfaces and colloids, 28(37), 13395-13404 (2012-08-16)
Fluorescence correlation spectroscopy (FCS) measurements are widely used for determination of diffusion coefficients of lipids and proteins in biological membranes. In recent years, several variants of FCS have been introduced. However, a comprehensive comparison of these methods on identical systems...
Zhixing Chen et al.
Journal of the American Chemical Society, 134(33), 13692-13699 (2012-08-10)
Chemical tags are now viable alternatives to fluorescent proteins for labeling proteins in living cells with organic fluorophores that have improved brightness and other specialized properties. Recently, we successfully rendered our TMP-tag covalent with a proximity-induced reaction between the protein...
John G Bruno et al.
Combinatorial chemistry & high throughput screening, 14(7), 622-630 (2011-05-04)
Several different approaches have been taken to development of homogeneous fluorescent aptamer assays including end-labeled beacons and signaling aptamers which are intrinsically quenched by nucleotides. Two new strategies dubbed "intrachain" and "competitive" FRET-aptamer assays are summarized in this review. Intrachain...
Sridharan Rajagopalan et al.
Nucleic acids research, 39(6), 2294-2303 (2010-11-26)
The state of oligomerization of the tumor suppressor p53 is an important factor in its various biological functions. It has a well-defined tetramerization domain, and the protein exists as monomers, dimers and tetramers in equilibrium. The dissociation constants between oligomeric...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.