74402

Sigma-Aldrich

Atto 488 iodoacetamide

NACRES:
NA.32

Quality Level

mol wt

Mw 913 g/mol

fluorescence

λex 501 nm; λem 523 nm

storage temp.

−20°C

General description

Atto 488 is a superior fluorescent label with high molecular absorption (90.000) and quantum yield (0.80) as well as sufficient stoke′s shift. It is optimized for excitation with argon laser, and is characterized by high photo stability.
The iodoacetamide derivative reacts, like the maleimide, with a sulfhydryl group forming a thioether bond. It is predominantly used for tagging cystein residues of proteins.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Monitoring single membrane protein dynamics in a liposome manipulated in solution by the ABELtrap.
Rendler, T., et al.
arXiv, 1102-1102 (2011)
Analysis of fluorescent nanostructures in biological systems by means of spectral position determination microscopy (SPDM).
Muller, P., et al. et al.
Current Microscopy Contributions to Advances in Science and Technology, 1, 3-12 (2012)
Jonas K Hannestad et al.
ACS nano, 7(1), 308-315 (2012-12-12)
We use single-molecule fluorescence microscopy to monitor individual hybridization reactions between membrane-anchored DNA strands, occurring in nanofluidic lipid monolayer films deposited on Teflon AF substrates. The DNA molecules are labeled with different fluorescent dyes, which make it possible to simultaneously...
Lucia De Rosa et al.
Organic & biomolecular chemistry, 10(2), 273-280 (2011-11-11)
In the last few years, the use of labeled proteins has significantly expanded in the life sciences. Now, labeled proteins are indispensable tools for a wide spectrum of biophysical and chemical biology applications. In particular, the quest for more sophisticated...
Tilak Jain et al.
Journal of structural biology, 179(1), 68-75 (2012-05-10)
Over the last three decades, Cryo-TEM has developed into a powerful technique for high-resolution imaging of biological macromolecules in their native vitrified state. However, the method for vitrifying specimens onto EM grids is essentially unchanged - application of ∼3 μL...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.