A1521

Sigma-Aldrich

β-Asp-Gly

Empirical Formula (Hill Notation):
C6H10N2O5
CAS Number:
Molecular Weight:
190.15
MDL number:
PubChem Substance ID:

storage temp.

−20°C

SMILES string

NC(CC(=O)NCC(O)=O)C(O)=O

InChI

1S/C6H10N2O5/c7-3(6(12)13)1-4(9)8-2-5(10)11/h3H,1-2,7H2,(H,8,9)(H,10,11)(H,12,13)

InChI key

ZTEDWFWBGPKUOD-UHFFFAOYSA-N

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
A E van den Bogaard et al.
The Journal of hygiene, 97(1), 49-59 (1986-08-01)
The impact of three current types of preoperative large bowel preparation on the microbial flora and the colonization resistance (CR) was investigated in 15 volunteers. In the first group a whole gut irrigation was performed without administration of antibiotics (group...
A V Lapchinskaia et al.
Antibiotiki i meditsinskaia biotekhnologiia = Antibiotics and medical biotechnology, 32(3), 206-210 (1987-03-01)
The effect of long-term use of cephalexin and erythromycin (for 17 days) on large intestine microflora was studied on rats. It was found that intragastric administration of cephalexin in a dose of 800 mg/kg and erythromycin in a dose of...
M Sato et al.
The Biochemical journal, 263(2), 617-620 (1989-10-15)
A novel o-phthalaldehyde-reactive compound was found in the h.p.l.c. chromatogram of Aplysia kurodai extract. This compound was isolated by ion-exchange chromatography and preparative high-voltage paper electrophoresis. It was shown by optical-rotatory-dispersion spectrum and optical-resolution h.p.l.c. analysis that this compound consisted...
H G de Vries-Hospers et al.
The Journal of infectious diseases, 150(5), 636-642 (1984-11-01)
Aztreonam, a minimally absorbable monobactam antibiotic that is active primarily against gram-negative bacilli, was given orally to 10 healthy volunteers in three regimens: 60 mg, 300 mg, and 1,500 mg daily for five days. The regimens were administered at least...
C E Leijonmarck et al.
Scandinavian journal of gastroenterology, 23(1), 59-64 (1988-01-01)
The effect of total parenteral nutrition (TPN) on the following six intestinal microflora-associated characteristics (MACs) was studied in patients with Crohn's disease: faecal tryptic activity (FTA), formation of coprostanol, urobilinogen, and deoxycholic acid, and degradation of mucin and beta-aspartylglycine. The...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.