MilliporeSigma
All Photos(1)

A1541

Sigma-Aldrich

L-Alanine 4-methoxy-β-naphthylamide

Empirical Formula (Hill Notation):
C14H16N2O2
CAS Number:
Molecular Weight:
244.29
MDL number:
PubChem Substance ID:

storage temp.

−20°C

SMILES string

COc1cc(NC(=O)[C@H](C)N)cc2ccccc12

Substrates

Fluorogenic substrate for aminopeptidase M

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service