MilliporeSigma
All Photos(1)

A1876

Sigma-Aldrich

N-Acetyl-DL-methionine magnesium salt

Linear Formula:
C14H24N2O6S2Mg
CAS Number:
Molecular Weight:
404.79
EC Number:
MDL number:

storage temp.

2-8°C

SMILES string

[Mg].CSCCC(NC(C)=O)C(O)=O

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service