MilliporeSigma
  • A284
All Photos(1)

A284

Sigma-Aldrich

PAK I Peptide Substrate

solid

Synonym(s):
AKRESAA, Ala-Lys-Arg-Glu-Ser-Ala-Ala
Empirical Formula (Hill Notation):
C29H53N11O11
Molecular Weight:
731.80

form

solid

color

white

storage temp.

−20°C

SMILES string

C[C@H](N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(O)=O

Substrates

Peptide substrate for γ-PAK (PAK I).

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service