MilliporeSigma
All Photos(1)

A3549

Sigma-Aldrich

Acetic anhydride-carbonyl-14C

~95%

Linear Formula:
[CH314CO]2O
CAS Number:
Molecular Weight:
102.09
MDL number:
PubChem Substance ID:

assay

~95%

extent of labeling

1-20 mCi per mmol

format

neat

mass shift

M+4

storage temp.

2-8°C

SMILES string

O[14C](=O)O[14C](O)=O

Looking for similar products? Visit Product Comparison Guide

Packaging

sealed ampule

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service