MilliporeSigma
All Photos(1)

A5055

Sigma-Aldrich

N-Acetyl-D-glucosaminitol

Synonym(s):
2-Acetamido-2-deoxy-D-glucitol
Empirical Formula (Hill Notation):
C8H17NO6
CAS Number:
Molecular Weight:
223.22
MDL number:

SMILES string

CC(=O)NC(CO)C(O)C(O)C(O)CO

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service