MilliporeSigma
All Photos(1)

A5553

Sigma-Aldrich

Bis(1-butylpentyl) adipate

Synonym(s):
Adipic acid di-5-nonyl ester, BBPA, Di(5-nonyl) adipate
Empirical Formula (Hill Notation):
C24H46O4
CAS Number:
Molecular Weight:
398.62
Beilstein:
6572484
EC Number:
MDL number:

density

0.910 g/mL at 20 °C (lit.)

SMILES string

CCCCC(CCCC)OC(=O)CCCCC(=O)OC(CCCC)CCCC

InChI

1S/C24H46O4/c1-5-9-15-21(16-10-6-2)27-23(25)19-13-14-20-24(26)28-22(17-11-7-3)18-12-8-4/h21-22H,5-20H2,1-4H3

InChI key

VXQKWLPFTKRXCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service