MilliporeSigma
All Photos(1)

A5667

Sigma-Aldrich

Asp-Ala-Glu-Asn-Leu-Ile-Asp-Ser-Phe-Gln-Glu-Ile-Val

≥97% (HPLC)

Synonym(s):
Prepro-gonadotropin releasing hormone fragment 14-26 human
Empirical Formula (Hill Notation):
C65H101N15O25
CAS Number:
Molecular Weight:
1492.58
MDL number:
PubChem Substance ID:

assay

≥97% (HPLC)

composition

Peptide content, ~65%

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](C)NC(=O)[C@@H](N)CC(O)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc1ccccc1)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](C(C)C)C(O)=O

InChI

1S/C65H101N15O25/c1-10-31(7)51(80-61(100)39(23-29(3)4)73-59(98)41(26-45(68)83)75-56(95)37(18-21-46(84)85)70-53(92)33(9)69-54(93)35(66)25-48(88)89)63(102)76-42(27-49(90)91)60(99)77-43(28-81)62(101)74-40(24-34-15-13-12-14-16-34)58(97)72-36(17-20-44(67)82)55(94)71-38(19-22-47(86)87)57(96)79-52(32(8)11-2)64(103)78-50(30(5)6)65(104)105/h12-16,29-33,35-43,50-52,81H,10-11,17-28,66H2,1-9H3,(H2,67,82)(H2,68,83)(H,69,93)(H,70,92)(H,71,94)(H,72,97)(H,73,98)(H,74,101)(H,75,95)(H,76,102)(H,77,99)(H,78,103)(H,79,96)(H,80,100)(H,84,85)(H,86,87)(H,88,89)(H,90,91)(H,104,105)/t31-,32-,33-,35-,36-,37-,38-,39-,40-,41-,42-,43-,50-,51-,52-/m0/s1

InChI key

SLLIDJFLMJLJIA-CXWSTQGDSA-N

Gene Information

human ... GNRH1(2796)

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service