MilliporeSigma
All Photos(1)

A7553

Sigma-Aldrich

L-Aspartic acid-UL-14C

aqueous ethanol solution

Linear Formula:
HO14CO14CH214CH[NH2]14COOH
CAS Number:
Molecular Weight:
133.10
MDL number:
PubChem Substance ID:

form

aqueous ethanol solution

extent of labeling

120-240 mCi per mmol

mass shift

M+8

storage temp.

2-8°C

SMILES string

N[14C@@H]([14CH2][14C](O)=O)[14C](O)=O

Packaging

serum bottle

Physical form

Aqueous solution containing 2% ethanol

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service