MilliporeSigma
All Photos(1)

A7760

Sigma-Aldrich

Arachidonyl alcohol

~99%, liquid

Empirical Formula (Hill Notation):
C20H34O
CAS Number:
Molecular Weight:
290.48
MDL number:

assay

~99%

form

liquid

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCCO

Packaging

Sealed ampule.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service