Documents

A9009

Sigma-Aldrich

D-Aspartic acid β-hydroxamate

Empirical Formula (Hill Notation):
C4H8N2O4
CAS Number:
Molecular Weight:
148.12

storage temp.

−20°C

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Xianfeng Gu et al.
Journal of medicinal chemistry, 49(25), 7532-7539 (2006-12-08)
Protein tyrosine kinases use two Mg(2+) ions as cofactors in catalysis, one as the ATP-Mg complex (M1) and the other as an essential activator (M2). The M2-binding site has high affinity for transition metal cations such as cobalt and zinc....
Roland Wakiec et al.
Antimicrobial agents and chemotherapy, 52(11), 4057-4063 (2008-09-17)
The susceptibility to several oligopeptide and amino acid antifungals of a Saccharomyces cerevisiae strain carrying multiple deletions in yeast multidrug resistance genes was compared to transformants containing the CDR1, CDR2, or MDR1 genes that encode the major Candida albicans drug...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.