MilliporeSigma
All Photos(1)

B1519

Sigma-Aldrich

Nα-Benzyl-L-histidine methyl ester dihydrochloride

Empirical Formula (Hill Notation):
C14H17N3O2 · 2HCl
CAS Number:
Molecular Weight:
332.23
MDL number:

storage temp.

−20°C

SMILES string

Cl.COC(=O)C(Cc1cnc[nH]1)NCc2ccccc2

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service