All Photos(1)

B3258

Sigma-Aldrich

N-Benzoyl-N-phenylhydroxylamine

Synonym(s):
N-Phenylbenzohydroxamic acid, N-Hydroxy-N-phenylbenzamide
Linear Formula:
C6H5CON(OH)C6H5
CAS Number:
Molecular Weight:
213.23
Beilstein:
2212449
EC Number:
MDL number:

mp

118-120 °C (lit.)

storage temp.

2-8°C

SMILES string

ON(c1ccccc1)C(=O)c2ccccc2

InChI

1S/C13H11NO2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,16H

InChI key

YLYIXDZITBMCIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service