MilliporeSigma
All Photos(1)

B4139

Sigma-Aldrich

Bovine adrenal medulla dodecapeptide

≥97% (HPLC)

Synonym(s):
BAM-12P, Tyr-Gly-Gly-Phe-Met-Arg-Arg-Val-Gly-Arg-Pro-Glu
Empirical Formula (Hill Notation):
C62H97N21O16S
CAS Number:
Molecular Weight:
1424.63
MDL number:
PubChem Substance ID:

assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(Cc1ccccc1)NC(=O)CNC(=O)CNC(=O)C(N)Cc2ccc(O)cc2)C(=O)NC(CCCNC(N)=N)C(=O)NC(CCCNC(N)=N)C(=O)NC(C(C)C)C(=O)NCC(=O)NC(CCCNC(N)=N)C(=O)N3CCCC3C(=O)NC(CCC(O)=O)C(O)=O

InChI

1S/C62H97N21O16S/c1-34(2)50(57(96)75-33-48(87)76-42(15-9-26-72-62(68)69)58(97)83-27-10-16-45(83)56(95)81-43(59(98)99)21-22-49(88)89)82-54(93)40(14-8-25-71-61(66)67)79-52(91)39(13-7-24-70-60(64)65)78-53(92)41(23-28-100-3)80-55(94)44(30-35-11-5-4-6-12-35)77-47(86)32-73-46(85)31-74-51(90)38(63)29-36-17-19-37(84)20-18-36/h4-6,11-12,17-20,34,38-45,50,84H,7-10,13-16,21-33,63H2,1-3H3,(H,73,85)(H,74,90)(H,75,96)(H,76,87)(H,77,86)(H,78,92)(H,79,91)(H,80,94)(H,81,95)(H,82,93)(H,88,89)(H,98,99)(H4,64,65,70)(H4,66,67,71)(H4,68,69,72)

InChI key

LNSYPNFYYCUMEW-UHFFFAOYSA-N

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

A C Camargo et al.
Biochemical and biophysical research communications, 130(2), 932-938 (1985-07-31)
The conversion of BAM-12P to Met-enkephalin and the hydrolysis of the Phe-Met and Phe-Leu bonds of met-enkephalin-Arg-Phe and Leu-enkephalin-Arg-Arg, respectively, by rabbit brain endo-oligopeptidase A were demonstrated. Peptide fragments were isolated by high performance liquid chromatography and identified by amino
N Tezapsidis et al.
FEBS letters, 246(1-2), 44-48 (1989-03-27)
An enzymatic activity has been identified which is capable of generating a product chromatographically identical with adrenorphin from the model substrate BAM12P. This enzyme was purified by gel filtration and ion-exchange chromatography and characterised as having a molecular mass between
A new endogenous opioid peptide from bovine adrenal medulla: isolation and amino acid sequence of a dodecapeptide (BAM-12P).
K Mizuno et al.
Biochemical and biophysical research communications, 95(4), 1482-1488 (1980-08-29)
A Baird et al.
Proceedings of the National Academy of Sciences of the United States of America, 79(6), 2023-2025 (1982-03-01)
A highly specific radioimmunoassay for one of the putative adrenomedullary [Met]enkephalin precursors, BAM-12P (Tyr-Gly-Gly-Phe-Met-Arg-Arg-Val-Gly-Arg-Pro-Glu-OH), has been developed. The BAM-12P antibodies recognize the COOH-terminal fragment of the peptide from Arg7 to Glu12 and do not crossreact with [Met5]- or [Leu5]enkephalin or
J L Maderdrut et al.
Brain research, 377(1), 29-40 (1986-07-02)
Met5-enkephalin- (Met-ENK), Leu5-enkephalin-, Met5-enkephalin-Arg6-Phe7-, metorphamide- and BAM 22P-like peptides could be detected in the lumbar spinal cord of the chicken by immunocytochemistry and/or high performance liquid chromatography. However, a peptide identical to Met5-enkephalin-Arg6-Gly7-Leu8 could not be detected in the lumbar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service