Documents

B4769

Sigma-Aldrich

Boc-D-Pro-OH

Synonym(s):
Boc-D-proline
Empirical Formula (Hill Notation):
C10H17NO4
CAS Number:
Molecular Weight:
215.25
Beilstein/REAXYS Number:
5744801
MDL number:

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)N1CCC[C@@H]1C(O)=O

Looking for similar products? Visit Product Comparison Guide

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.