MilliporeSigma
All Photos(1)

B4791

Sigma-Aldrich

[des-Pro3, Ala2,6]-Bradykinin

≥97% (HPLC)

Empirical Formula (Hill Notation):
C43H64N14O9
CAS Number:
Molecular Weight:
921.06
MDL number:
PubChem Substance ID:

assay

≥97% (HPLC)

composition

Peptide content, ~70%

storage temp.

−20°C

SMILES string

CC(NC(=O)C(N)CCCNC(N)=N)C(=O)NCC(=O)NC(Cc1ccccc1)C(=O)NC(C)C(=O)N2CCCC2C(=O)NC(Cc3ccccc3)C(=O)NC(CCCNC(N)=N)C(O)=O

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

D Chaturvedi et al.
Peptide research, 6(6), 308-312 (1993-11-01)
A series of peptide analogs and fragments of bradykinin were designed and synthesized on solid supports using Boc and Fmoc strategies, and on polyethylene pins using Fmoc strategy. The peptides were purified, characterized and tested for their inhibitory effects on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service