MilliporeSigma
All Photos(1)

B7754

Sigma-Aldrich

Boc-L-Histidine hydrazide

Empirical Formula (Hill Notation):
C11H19N5O3
CAS Number:
Molecular Weight:
269.30
EC Number:
MDL number:
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)NC(Cc1cnc[nH]1)C(=O)NN

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service