B7936

Sigma-Aldrich

BBMP

≥98% (HPLC), solid

Synonym(s):
5-(Benylsulfonyl)-4-bromo-2-methyl-3(2H)-pyridazinone
Empirical Formula (Hill Notation):
C12H11BrN2O3S
CAS Number:
Molecular Weight:
343.20
MDL number:

assay

≥98% (HPLC)

form

solid

solubility

DMSO: 16 mg/mL
H2O: <2 mg/mL

storage temp.

2-8°C

SMILES string

CN1N=CC(=C(Br)C1=O)S(=O)(=O)Cc2ccccc2

Biochem/physiol Actions

BBMP is a mitochondrial permeability transition pore (PTP) inhibitor. PTP inhibitors and mitochondrial depolarization are potential therapeutics for neurodegenerative diseases.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service