B9382

Sigma-Aldrich

N-Benzoyl-L-phenylalanine

Empirical Formula (Hill Notation):
C16H15NO3
CAS Number:
Molecular Weight:
269.30
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

storage temp.

−20°C

SMILES string

OC(=O)C(Cc1ccccc1)NC(=O)c2ccccc2

InChI

1S/C16H15NO3/c18-15(13-9-5-2-6-10-13)17-14(16(19)20)11-12-7-3-1-4-8-12/h1-10,14H,11H2,(H,17,18)(H,19,20)

InChI key

NPKISZUVEBESJI-UHFFFAOYSA-N

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Leelavinothan Pari et al.
Therapie, 61(2), 125-131 (2006-08-05)
The effect of N-benzoyl-D-phenylalanine (NBDP) and metformin on neonatal streptozotocin (nSTZ) induced diabetes has been studied on plasma and tissue glycoproteins. In some pathological conditions, such as cancer, rheumatoid arthritis and diabetes, there is an abnormal glycosylation of acute phase...
Leelavinothan Pari et al.
Pharmacological reports : PR, 57(4), 498-503 (2005-09-01)
The present investigation was undertaken to study the effect of treatment with D-phenylalanine derivative and metformin in neonatal streptozotocin (nSTZ)-induced non-insulin-dependent diabetes mellitus (NIDDM) in rats. To induce NIDDM, a single dose injection of streptozotozin (STZ) (100 mg kg(-1); ip)...
Emanuela Occhipinti et al.
Biochimica et biophysica acta, 1764(3), 563-572 (2006-02-01)
Sulfolobus solfataricus carboxypeptidase, (CPSso), is a heat- and pressure-resistant zinc-metalloprotease. Thanks to its properties, it is an ideal tool for investigating the role of non-covalent interactions in substrate binding. It has a broad substrate specificity as it can cleave any...
N Ashokkumar et al.
The Journal of pharmacy and pharmacology, 57(3), 359-366 (2005-04-06)
The effect of N-benzoyl-D-phenylalanine (NBDP) and metformin combination treatment on circulatory lipids, lipoproteins and lipid peroxidation markers were studied in neonatal streptozotocin (nSTZ) non-insulin dependent diabetic rats. Non-insulin dependent diabetes mellitus (NIDDM) was induced by a single dose injection of...
Christopher W Lennon et al.
Genes & development, 26(23), 2634-2646 (2012-12-05)
Escherichia coli DksA is a transcription factor that binds to RNA polymerase (RNAP) without binding to DNA, destabilizing RNAP-promoter interactions, sensitizing RNAP to the global regulator ppGpp, and regulating transcription of several hundred target genes, including those encoding rRNA. Previously...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.