MilliporeSigma
All Photos(1)

C1125

Sigma-Aldrich

Calcium phosphate

gel (aged)

CAS Number:
MDL number:
PubChem Substance ID:

form

gel (aged)

composition

Solids content, 5-20%

storage temp.

2-8°C

SMILES string

O.O.[Ca++].OP([O-])([O-])=O

InChI

1S/Ca.H3O4P.2H2O/c;1-5(2,3)4;;/h;(H3,1,2,3,4);2*1H2/q+2;;;/p-2

InChI key

XAAHAAMILDNBPS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Quality

Purified of extraneous salts.

Caution

Do Not Freeze. Inspect all shipments promptly on arrival for loss of gel structure caused by freezing en route.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service