MilliporeSigma
All Photos(1)

C1410

Sigma-Aldrich

Cibacron Blue 3GA Agarose

Type 3000-L, lyophilized powder

MDL number:
PubChem Substance ID:

type

Type 3000-L

form

lyophilized powder

extent of labeling

2-5 μmol per mL

matrix

4% beaded agarose

capacity

8-15 mg/mL binding capacity (bovine serum albumin)

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].Nc1c(cc(Nc2ccc(Nc3nc(Cl)nc(Nc4cccc(c4)S([O-])(=O)=O)n3)c(c2)S([O-])(=O)=O)c5C(=O)c6ccccc6C(=O)c15)S([O-])(=O)=O

InChI

1S/C29H20ClN7O11S3.3Na/c30-27-35-28(33-13-4-3-5-15(10-13)49(40,41)42)37-29(36-27)34-18-9-8-14(11-20(18)50(43,44)45)32-19-12-21(51(46,47)48)24(31)23-22(19)25(38)16-6-1-2-7-17(16)26(23)39;;;/h1-12,32H,31H2,(H,40,41,42)(H,43,44,45)(H,46,47,48)(H2,33,34,35,36,37);;;/q;3*+1/p-3

InChI key

VZPXDCIISFTYOM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Quantity

Swelling: 1 g swells to approx. 5 ml

Physical form

Stabilized with lactose

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Product Information Sheet

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service