All Photos(1)

C1894

Sigma-Aldrich

Z-Asp(OBzl)-ONp

Synonym(s):
Z-L-aspartic acid β-benzyl α-(4-nitrophenyl) ester, Z-L-aspartic acid 4-benzyl 1-(4-nitrophenyl) ester
Empirical Formula (Hill Notation):
C25H22N2O8
CAS Number:
Molecular Weight:
478.45
Beilstein:
1898340
MDL number:

storage temp.

−20°C

SMILES string

[O-][N+](=O)c1ccc(OC(=O)[C@H](CC(=O)OCc2ccccc2)NC(=O)OCc3ccccc3)cc1

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service