C2110

Sigma-Aldrich

Arrayer Calibration Solution

Pricing and availability is not currently available.

foreign activity

DNase, RNase, free

storage temp.

2-8°C

General description

Ready-to-use solution for calibration of arrayers prior to printing microarrays. Useful in determining that each pin of the arrayer is printing spots of equal size and uniform morphology. For use with split-pin and pin-ring arrayers.

WGK Germany

-

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.