MilliporeSigma
All Photos(1)

C2921

Sigma-Aldrich

Corazonin

≥97% (HPLC)

Synonym(s):
pGlu-Thr-Phe-Gln-Tyr-Ser-Arg-Gly-Trp-Thr-Asn-NH2
Empirical Formula (Hill Notation):
C62H84N18O18
CAS Number:
Molecular Weight:
1369.44
MDL number:
PubChem Substance ID:

assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(O)C(NC(=O)C(Cc1c[nH]c2ccccc12)NC(=O)CNC(=O)C(CCCNC(N)=N)NC(=O)C(CO)NC(=O)C(Cc3ccc(O)cc3)NC(=O)C(CCC(N)=O)NC(=O)C(Cc4ccccc4)NC(=O)C(NC(=O)C5CCC(=O)N5)C(C)O)C(=O)NC(CC(N)=O)C(N)=O

Biochem/physiol Actions

Cardioaccelerating peptide originally isolated from the corpus cardiacum of the American cockroach; the most potent insect cardioactive neuropeptide.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

J A Veenstra
FEBS letters, 250(2), 231-234 (1989-07-03)
Corazonin, a new cardioaccelerating peptide, has been isolated from the corpora cardiaca of the American cockroach, Periplaneta americana, and its structure determined to be Glp-Thr-Phe-Gln-Tyr-Ser-Arg-Gly-Trp-Thr-Asn-amide. The peptide stimulated heart beat at concentrations as low as 0.2 nM, which makes it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service