MilliporeSigma
All Photos(1)

C3910

Sigma-Aldrich

(2R,3R)-3-(Z-amino)-2-hydroxy-4-phenylbutyric acid

Synonym(s):
N-Cbz-(2R,3R)-3-amino-2-hydroxy-4-phenylbutyric acid
Empirical Formula (Hill Notation):
C18H19NO5
CAS Number:
Molecular Weight:
329.35
MDL number:
PubChem Substance ID:

storage temp.

−20°C

SMILES string

[H][C@](Cc1ccccc1)(NC(=O)OCc2ccccc2)[C@@H](O)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Preparation Note

CBZ-protected novel amino acid in epibestatin.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service