MilliporeSigma
All Photos(1)

C5627

Sigma-Aldrich

Z-Tyr-OH

Synonym(s):
N-Carbobenzyloxy-L-tyrosine, Z-L-Tyrosine
Linear Formula:
HOC6H4CH2CH(NHCOOCH2C6H5)COOH
CAS Number:
Molecular Weight:
315.32
Beilstein:
2169918
EC Number:
MDL number:

mp

57-60 °C (lit.)

storage temp.

−20°C

SMILES string

O=C(N[C@H](C(O)=O)CC1=CC=C(O)C=C1)OCC2=CC=CC=C2

InChI

1S/C17H17NO5.H2O/c19-14-8-6-12(7-9-14)10-15(16(20)21)18-17(22)23-11-13-4-2-1-3-5-13;/h1-9,15,19H,10-11H2,(H,18,22)(H,20,21);1H2/t15-;/m0./s1

InChI key

BBPCQKGWAVVMSL-RSAXXLAASA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service