MilliporeSigma
All Photos(1)

C8321

Sigma-Aldrich

Cibacron Blue 3GA Agarose

saline suspension, Fast flow

MDL number:
PubChem Substance ID:

form

saline suspension

extent of labeling

≥6 μmol per mL

matrix

Fast flow highly cross-linked 6% agarose

matrix attachment

triazine coupling

capacity

≥15 mg/mL binding capacity (human serum albumin)

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].Nc1c(cc(Nc2ccc(Nc3nc(Cl)nc(Nc4cccc(c4)S([O-])(=O)=O)n3)c(c2)S([O-])(=O)=O)c5C(=O)c6ccccc6C(=O)c15)S([O-])(=O)=O

InChI

1S/C29H20ClN7O11S3.3Na/c30-27-35-28(33-13-4-3-5-15(10-13)49(40,41)42)37-29(36-27)34-18-9-8-14(11-20(18)50(43,44)45)32-19-12-21(51(46,47)48)24(31)23-22(19)25(38)16-6-1-2-7-17(16)26(23)39;;;/h1-12,32H,31H2,(H,40,41,42)(H,43,44,45)(H,46,47,48)(H2,33,34,35,36,37);;;/q;3*+1/p-3

InChI key

VZPXDCIISFTYOM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Cibacron Blue 3GA-agarose is an agarose conjugate in saline suspension used in affinity chromatography, protein chromatography and dye resins.

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Product Information Sheet

More documents

Quotes and Ordering

María del Pilar Ferraris et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(26), 2741-2745 (2011-08-30)
The objective of this study was the development of affinity adsorbent particles with the appropriate characteristics to be applied in protein purification using the affinity ultrafiltration method. To prepare affinity macroligands Cibacron Blue 3GA, as a ligand molecule, was immobilized
Richa Gupta et al.
Molecular biology reports, 39(5), 5607-5614 (2011-12-23)
MspNI and MspNII, isoschizomers of prototype Type II restriction endonucleases AvaII and BstYI, were extracted from an extreme thermophile bacterium belonging to the genus Meiothermus, isolated from the hot sulphur springs in north Himalayan region of India where temperature and
Takashi Miura et al.
Free radical biology & medicine, 51(11), 2082-2089 (2011-10-04)
1,2-Naphthoquinone (1,2-NQ) is electrophilic, and forms covalent bonds with protein thiols, but its two-electron reduction product 1,2-dihydroxynaphthalene (1,2-NQH(2)) is not, so enzymes catalyzing the reduction with reduced pyridine nucleotides as cofactors could protect cells from electrophile-based chemical insults. To assess
Christa E Müller et al.
Bioorganic & medicinal chemistry letters, 16(23), 5943-5947 (2006-09-26)
Polyoxotungstates were identified as potent inhibitors of NTPDases1, 2, and 3. The most potent compound was K(6)H(2)[TiW(11)CoO(40)], exhibiting K(i) values of 0.140 microM (NTPDase1), 0.910 microM (NTPDase2), and 0.563 microM (NTPDase3). One of the compounds, (NH(4))(18)[NaSb(9)W(21)O(86)], was selective for NTPDases2
Younis Baqi et al.
Journal of medicinal chemistry, 53(5), 2076-2086 (2010-02-12)
ecto-5'-Nucleotidase (eN, CD73) plays a major role in controlling extracellular adenosine levels. eN inhibitors have potential as novel drugs, for example, for the treatment of cancer. In the present study, we synthesized and investigated a series of 55 anthraquinone derivatives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service