All Photos(1)

D0880

Sigma-Aldrich

DNP-DL-norleucine

Synonym(s):
N-(2,4-Dinitrophenyl)-DL-norleucine
Empirical Formula (Hill Notation):
C12H15N3O6
CAS Number:
Molecular Weight:
297.26
EC Number:
MDL number:
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CCCCC(Nc1ccc(cc1N(=O)=O)N(=O)=O)C(O)=O

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service